Assay Details
| Gene Symbol: Lnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077429.1 | NP_001070897.1 | 6 - 7 | 479 | 214 | |
| XM_006234377.2 | XP_006234439.1 | 6 - 7 | 561 | 214 | ||
| XM_006234378.2 | XP_006234440.1 | 7 - 8 | 582 | 214 | ||
| XM_006234379.2 | XP_006234441.1 | 6 - 7 | 609 | 214 | ||
| XM_006234380.2 | XP_006234442.1 | 6 - 7 | 681 | 214 | ||
| XM_006234381.2 | XP_006234443.1 | 6 - 7 | 774 | 214 | ||
| XM_006234382.2 | XP_006234444.1 | 7 - 8 | 702 | 214 | ||
| XM_006234385.2 | XP_006234447.1 | 6 - 7 | 563 | 214 | ||
| XM_008761909.1 | XP_008760131.1 | 6 - 7 | 1658 | 214 | ||
| XM_008761910.1 | XP_008760132.1 | 6 - 7 | 563 | 214 | ||
| GenBank mRNA | BC126100.1 | - | 6 - 7 | 479 | 7132654 | 214 |
| FQ218047.1 | - | 5 - 6 | 358 | 214 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lnp
Genomic Map