Assay Details
Gene Symbol: Icmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133310.2 | NP_579844.2 | 3 - 4 | 506 | 68 | |
GenBank mRNA | AF075595.1 | - | 3 - 4 | 294 | 68 | |
BC083923.1 | - | 4 - 5 | 711 | 68 | ||
CB616023.1 | - | 3 - 4 | 506 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
