Assay Details
| Gene Symbol: LOC681419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037919.1 | 2 - 3 | 234 | 74 | ||
| GenBank mRNA | EV762401.1 | - | 2 - 3 | 234 | 74 | |
| Gene Symbol: Rbbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107912.2 | NP_001101382.1 | 5 - 6 | 610 | 74 | |
| GenBank mRNA | BC168994.1 | - | 5 - 6 | 610 | 8362559 | 74 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC681419
Genomic Map