Assay Details
| Gene Symbol: Sptlc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037097.1 | NP_001032174.1 | 8 - 9 | 1270 | 66 | |
| XM_008764823.1 | XP_008763045.1 | 8 - 9 | 1365 | 66 | ||
| GenBank mRNA | BC107662.1 | - | 8 - 9 | 1270 | 7936642 | 66 |
More Information
Updated Mapping Information
Set Membership: |
|

Sptlc2
Genomic Map