Assay Details
| Gene Symbol: Flvcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199109.2 | NP_954540.2 | 2 - 3 | 1153 | 63 | |
| GenBank mRNA | AY260573.1 | - | 2 - 3 | 905 | 63 | |
| BC101899.1 | - | 2 - 3 | 1153 | 7932187 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Flvcr2
Genomic Map