Assay Details
| Gene Symbol: Flvcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199109.2 | NP_954540.2 | 9 - 10 | 1858 | 63 | |
| GenBank mRNA | AY260573.1 | - | 9 - 10 | 1610 | 63 | |
| BC101899.1 | - | 9 - 10 | 1858 | 7932187 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P025658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129842 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P025658 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P025658 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P025658 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P320737 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26518 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40347 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59076 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51357 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73197 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98975 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 2512 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10886059 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886051 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886052 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886054 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886061 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886050 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886058 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886057 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886056 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886055 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886060 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886053 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10886049 | - | - |
| Rat Genome 230 2.0 Array | 1376036_at | - | - |
| Rat Genome 230 2.0 Array | 1386205_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377027_at | - | - |
| Rat Genome 230 2.0 Array | 1377026_a_at | - | - |
| Rat Genome 230 2.0 Array | 1376036_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1386205_at | - | - |
| Rat Genome 230 2.0 Array | 1377027_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377026_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1376036_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376036_at | - | - |
| Affymetrix HT Rat Focus Array | 1386205_at | - | - |
| Affymetrix HT Rat Focus Array | 1386205_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600024479.rn.1 | TC0600003485.rn.1 | RTA TC |
| Rat Expression Set 230 | 1386205_at | - | - |
| Rat Expression Set 230 | 1376036_at | - | - |
| Rat Expression Set 230 | 1386205_PM_at | - | - |
| Rat Expression Set 230 | 1376036_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10886049 | - | - |
| Rat Genome U34 Set | rc_AA956782_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21537046 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361794 | - | - |
| RatRef-12 Expression BeadChip | ILMN_63433 | - | - |

Flvcr2
Genomic Map