Assay Details
| Gene Symbol: Ttc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134731.1 | NP_001128203.1 | 2 - 3 | 662 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P083474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71921 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083474 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083474 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P083474 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P473622 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10885696 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885694 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885695 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10885693 | - | - |
| Rat Genome 230 2.0 Array | 1376013_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392632_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1376013_at | - | - |
| Rat Genome 230 2.0 Array | 1392632_at | - | - |
| Affymetrix HT Rat Focus Array | 1392632_at | - | - |
| Affymetrix HT Rat Focus Array | 1376013_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392632_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376013_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600007699.rn.1 | TC0600001115.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600007700.rn.1 | TC0600001115.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600007698.rn.1 | TC0600001115.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376013_at | - | - |
| Rat Expression Set 230 | 1376013_PM_at | - | - |
| Rat Expression Set 230 | 1392632_PM_at | - | - |
| Rat Expression Set 230 | 1392632_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10885693 | - | - |
| Rat Genome U34 Set | rc_AA925302_at | - | - |
| Rat Genome U34 Set | rc_AI044816_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21275196 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369250 | - | - |

Ttc9
Genomic Map