Assay Details
| Gene Symbol: Ttc9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134731.1 | NP_001128203.1 | 1 - 2 | 483 | 86 | |
| GenBank mRNA | BC166802.1 | - | 1 - 2 | 409 | 7938663 | 86 |
More Information
Updated Mapping Information
Set Membership: |
|

Ttc9
Genomic Map