Assay Details
| Gene Symbol: Plekhn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134523.1 | NP_001127995.1 | 2 - 3 | 746 | 97 | |
| XM_006239564.2 | XP_006239626.1 | 2 - 3 | 977 | 97 | ||
| XM_006239565.2 | XP_006239627.1 | 2 - 3 | 977 | 97 | ||
| XM_006239566.2 | XP_006239628.1 | 2 - 3 | 977 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plekhn1
Genomic Map