Assay Details
| Gene Symbol: Ube4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271198.1 | NP_001258127.1 | 8 - 9 | 1894 | 55 | |
| XM_006239387.2 | XP_006239449.1 | 10 - 11 | 2428 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ube4b
Genomic Map