Assay Details
| Gene Symbol: Ctrc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077649.2 | NP_001071117.1 | 3 - 4 | 236 | 59 | |
| GenBank mRNA | S80379.1 | - | 3 - 4 | 237 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctrc
Genomic Map