Assay Details
| Gene Symbol: Matn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006979.1 | NP_001006980.1 | 6 - 7 | 1382 | 68 | |
| GenBank mRNA | BC083869.1 | - | 6 - 7 | 1382 | 7128388 | 68 |
More Information
Updated Mapping Information
Set Membership: |
|

Matn1
Genomic Map