Assay Details
| Gene Symbol: Pramef12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239346.2 | XP_006239408.1 | 2 - 3 | 408 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pramef12| Gene Symbol: Pramef12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006239346.2 | XP_006239408.1 | 2 - 3 | 408 | 60 | |
Set Membership: |
|