Assay Details
| Gene Symbol: Ahdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134956.1 | NP_001128428.1 | 3 - 4 | 494 | 63 | |
| XM_008764151.1 | XP_008762373.1 | 1 - 2 | 220 | 63 | ||
| XM_008764152.1 | XP_008762374.1 | 1 - 2 | 115 | 63 | ||
| XM_008764153.1 | XP_008762375.1 | 1 - 2 | 234 | 63 | ||
| XM_008764154.1 | XP_008762376.1 | 1 - 2 | 234 | 63 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001134956.1,XM_008764151.1,XM_008764152.1,XM_008764153.1,XM_008764154.1 transcripts.
Set Membership: |
|

Ahdc1
Genomic Map