Assay Details
| Gene Symbol: Ahdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134956.1 | NP_001128428.1 | 5 - 6 | 5436 | 150 | |
| XM_008764151.1 | XP_008762373.1 | 3 - 4 | 5162 | 150 | ||
| XM_008764152.1 | XP_008762374.1 | 3 - 4 | 5057 | 150 | ||
| XM_008764153.1 | XP_008762375.1 | 3 - 4 | 5200 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P098931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348218 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167047 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P098931 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P098931 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P098931 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1042859 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Rat Genome 230 2.0 Array | 1391419_at | - | - |
| Rat Genome 230 2.0 Array | 1391419_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391419_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391419_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500011621.rn.1 | TC0500001614.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500011619.rn.1 | TC0500001614.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500011622.rn.1 | TC0500001614.rn.1 | RTA TC |
| Rat Expression Set 230 | 1391419_PM_at | - | - |
| Rat Expression Set 230 | 1391419_at | - | - |
| Rat Genome U34 Set | rc_AI102440_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21323547 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353314 | - | - |

Ahdc1
Genomic Map