Assay Details
| Gene Symbol: Tmem44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768813.1 | XP_008767035.1 | 6 - 7 | 1958 | 97 | |
| XM_213600.8 | XP_213600.7 | 6 - 7 | 1958 | 97 | ||
More Information
Set Membership: |
|

Tmem44
Genomic Map