Assay Details
| Gene Symbol: Lsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013421.1 | NP_001013439.1 | 13 - 14 | 1804 | 101 | |
| GenBank mRNA | BC079013.1 | - | 12 - 13 | 1650 | 101 | |
| BC091338.1 | - | 13 - 14 | 1804 | 7315079 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P525098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882545 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325867 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92928 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33555 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P525098 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P525098 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P525098 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P956938 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P525098 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1040 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91038 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80170 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10751743 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751754 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751742 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751753 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751750 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751756 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751751 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751755 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751744 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751745 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751749 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751748 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751741 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751746 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751747 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751752 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10751740 | - | - |
| Rat Genome 230 2.0 Array | 1391565_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391565_at | - | - |
| Rat Genome 230 2.0 Array | 1373293_at | - | - |
| Rat Genome 230 2.0 Array | 1373293_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391565_at | - | - |
| Affymetrix HT Rat Focus Array | 1391565_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373293_at | - | - |
| Affymetrix HT Rat Focus Array | 1373293_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100004411.rn.1 | TC1100000798.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100004414.rn.1 | TC1100000798.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100004413.rn.1 | TC1100000798.rn.1 | RTA TC |
| Rat Expression Set 230 | 1373293_PM_at | - | - |
| Rat Expression Set 230 | 1373293_at | - | - |
| Rat Expression Set 230 | 1391565_PM_at | - | - |
| Rat Expression Set 230 | 1391565_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10751740 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22203139 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365589 | - | - |

Lsg1
Genomic Map