Assay Details
| Gene Symbol: Zgrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233293.2 | XP_006233355.1 | 11 - 12 | 4854 | 70 | |
| XM_006233294.2 | XP_006233356.1 | 11 - 12 | 4250 | 70 | ||
| XM_006233296.2 | XP_006233358.1 | 9 - 10 | 4540 | 70 | ||
| XM_006233297.2 | XP_006233359.1 | 6 - 7 | 3343 | 70 | ||
| XM_008761479.1 | XP_008759701.1 | 10 - 11 | 4745 | 70 | ||
| XM_008761480.1 | XP_008759702.1 | 10 - 11 | 4167 | 70 | ||
More Information
Set Membership: |
|

Zgrf1
Genomic Map