Assay Details
| Gene Symbol: Palmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025688.1 | NP_001020859.1 | 769 | 126 | ||
| GenBank mRNA | BC098755.1 | - | 769 | 7370232 | 126 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P423490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420952 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913241 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31113 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P423490 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P423490 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P423490 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P423490 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 28625 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 62679 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 61983 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10826374 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826378 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826373 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826377 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826375 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826376 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826372 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826379 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10826371 | - | - |
| Rat Genome 230 2.0 Array | 1382864_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1376925_at | - | - |
| Rat Genome 230 2.0 Array | 1382864_at | - | - |
| Rat Genome 230 2.0 Array | 1376924_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1376925_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1376924_a_at | - | - |
| Rat Genome 230 2.0 Array | 1395734_at | - | - |
| Rat Genome 230 2.0 Array | 1395734_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376925_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376925_at | - | - |
| Affymetrix HT Rat Focus Array | 1382864_at | - | - |
| Affymetrix HT Rat Focus Array | 1376924_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1395734_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395734_at | - | - |
| Affymetrix HT Rat Focus Array | 1382864_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376924_a_at | - | - |
| Rat Expression Set 230 | 1382864_PM_at | - | - |
| Rat Expression Set 230 | 1376924_a_at | - | - |
| Rat Expression Set 230 | 1376924_PM_a_at | - | - |
| Rat Expression Set 230 | 1382864_at | - | - |
| Rat Expression Set 230 | 1376925_at | - | - |
| Rat Expression Set 230 | 1376925_PM_at | - | - |
| Rat Expression Set 230 | 1395734_at | - | - |
| Rat Expression Set 230 | 1395734_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10826371 | - | - |
| Rat Genome U34 Set | rc_AI231808_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21569506 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350622 | - | - |

Palmd
Genomic Map