Assay Details
| Gene Symbol: Syde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749414.3 | XP_003749462.1 | 4 - 5 | 3770 | 100 | |
| GenBank mRNA | FQ218152.1 | - | 2 - 3 | 275 | 100 | |
More Information
Set Membership: |
|

Syde2
Genomic Map