Assay Details
| Gene Symbol: Syde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749414.3 | XP_003749462.1 | 6 - 7 | 4181 | 95 | |
| GenBank mRNA | FQ218152.1 | - | 4 - 5 | 686 | 95 | |
More Information
Set Membership: |
|

Syde2
Genomic Map