Assay Details
| Gene Symbol: Stpg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100993.1 | NP_001094463.1 | 2 - 3 | 428 | 58 | |
| XM_008761520.1 | XP_008759742.1 | 2 - 3 | 522 | 58 | ||
| GenBank mRNA | BC128777.1 | - | 2 - 3 | 428 | 7134244 | 58 |
More Information
Updated Mapping Information
Set Membership: |
|

Stpg2
Genomic Map