Assay Details
| Gene Symbol: Peg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CB711252.1 | - | 120 | 75 | ||
| CB743994.1 | - | 76 | ||||
| CB766724.1 | - | 179 | 76 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Peg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CB711252.1 | - | 120 | 75 | ||
| CB743994.1 | - | 76 | ||||
| CB766724.1 | - | 179 | 76 | |||
Set Membership: |
|