Assay Details
| Gene Symbol: Dym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106133.2 | NP_001099603.2 | 4 - 5 | 546 | 62 | |
| XM_008772103.1 | XP_008770325.1 | 4 - 5 | 611 | 62 | ||
| XM_008772104.1 | XP_008770326.1 | 4 - 5 | 611 | 62 | ||
| GenBank mRNA | BC168151.1 | - | 4 - 5 | 522 | 7459379 | 62 |
More Information
Updated Mapping Information
Set Membership: |
|

Dym
Genomic Map