Assay Details
| Gene Symbol: Cdh10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168631.1 | NP_001162102.1 | 3 - 4 | 777 | 71 | |
| XM_006232079.2 | XP_006232141.1 | 3 - 4 | 853 | 71 | ||
| XM_006232080.2 | XP_006232142.1 | 3 - 4 | 885 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh10
Genomic Map