Assay Details
| Gene Symbol: Dap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022526.2 | NP_071971.1 | 3 - 4 | 388 | 88 | |
| GenBank mRNA | BC060569.1 | - | 3 - 4 | 206 | 6890457 | 88 |
| FQ218203.1 | - | 3 - 4 | 273 | 88 | ||
| FQ228698.1 | - | 3 - 4 | 226 | 88 | ||
| FQ228865.1 | - | 3 - 4 | 232 | 88 | ||
| U05334.1 | - | 3 - 4 | 389 | 88 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
autophagy
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of autophagy
negative regulation of NF-kappaB transcription factor activity
cellular response to amino acid starvation
negative regulation of transcription, DNA-templated
apoptotic signaling pathway
death domain binding
autophagy
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of autophagy
negative regulation of NF-kappaB transcription factor activity
cellular response to amino acid starvation
negative regulation of transcription, DNA-templated
apoptotic signaling pathway
death domain binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P099923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229769 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569442 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175281 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P099923 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P099923 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P099923 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P306639 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700485510 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46558 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29426 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29425 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700485510 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10814102 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814100 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814103 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814104 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814101 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814099 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10814098 | - | - |
| Rat Genome 230 2.0 Array | 1395881_at | - | - |
| Rat Genome 230 2.0 Array | 1395881_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369941_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369941_at | - | - |
| Affymetrix HT Rat Focus Array | 1395881_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369941_at | - | - |
| Affymetrix HT Rat Focus Array | 1369941_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1395881_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200004399.rn.1 | TC0200000713.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200004398.rn.1 | TC0200000713.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200004401.rn.1 | TC0200000713.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200004402.rn.1 | TC0200000713.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369941_PM_at | - | - |
| Rat Expression Set 230 | 1369941_at | - | - |
| Rat Expression Set 230 | 1395881_at | - | - |
| Rat Expression Set 230 | 1395881_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814098 | - | - |
| Rat Genome U34 Set | rc_AA851650_s_at | - | - |
| Rat Genome U34 Set | rc_AI178788_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22389695 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362377 | - | - |
| RatRef-12 Expression BeadChip | ILMN_64501 | - | - |

Dap
Genomic Map