Assay Details
| Gene Symbol: Tmem45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033067.1 | NP_001028239.1 | 1 - 2 | 215 | 70 | |
| GenBank mRNA | BC100635.1 | - | 1 - 2 | 215 | 5621332 | 70 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC100635.1,NM_001033067.1 transcripts.
Set Membership: |
|

Tmem45b
Genomic Map