Assay Details
| Gene Symbol: Tmem45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033067.1 | NP_001028239.1 | 3 - 4 | 624 | 62 | |
| GenBank mRNA | BC100635.1 | - | 3 - 4 | 624 | 5621332 | 62 |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem45b
Genomic Map