Assay Details
| Gene Symbol: Tmem45b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033067.1 | NP_001028239.1 | 5 - 6 | 948 | 64 | |
| GenBank mRNA | BC100635.1 | - | 5 - 6 | 948 | 5621332 | 64 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P710192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553515 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144769 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76659 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P710192 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P710192 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P710192 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P823351 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P710192 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58380 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19626 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 25458 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13265 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10915972 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10915974 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10915971 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10915975 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10915973 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10915976 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10915970 | - | - |
| Rat Genome 230 2.0 Array | 1390123_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1390123_at | - | - |
| Affymetrix HT Rat Focus Array | 1390123_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1390123_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800017622.rn.1 | TC0800002182.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800017623.rn.1 | TC0800002182.rn.1 | RTA TC |
| Rat Expression Set 230 | 1390123_PM_at | - | - |
| Rat Expression Set 230 | 1390123_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10915970 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21299615 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370988 | - | - |

Tmem45b
Genomic Map