Assay Details
| Gene Symbol: Acat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006995.1 | NP_001006996.1 | 2 - 3 | 214 | 63 | |
| GenBank mRNA | AY325187.1 | - | 188 | 63 | ||
| BC083872.1 | - | 2 - 3 | 214 | 7128564 | 63 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P024620 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929027 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152677 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P024620 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P131224 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P024620 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P131224 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P024620 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P131224 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P271586 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16774 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16724 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 674 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 79633 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10702948 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702958 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702964 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702946 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702952 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702962 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702947 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702949 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702950 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702951 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702959 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702960 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702953 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702961 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702954 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10702955 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10702945 | - | - |
| Rat Genome 230 2.0 Array | 1379249_at | - | - |
| Rat Genome 230 2.0 Array | 1379249_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372462_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372462_at | - | - |
| Affymetrix HT Rat Focus Array | 1372462_at | - | - |
| Affymetrix HT Rat Focus Array | 1372462_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100003138.rn.1 | TC0100000503.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003143.rn.1 | TC0100000503.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003142.rn.1 | TC0100000503.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003147.rn.1 | TC0100000503.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003148.rn.1 | TC0100000503.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003141.rn.1 | TC0100000503.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003140.rn.1 | TC0100000503.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003149.rn.1 | TC0100000503.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003144.rn.1 | TC0100000503.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100003151.rn.1 | TC0100000503.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372462_PM_at | - | - |
| Rat Expression Set 230 | 1372462_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702945 | - | - |
| Rat Genome U34 Set | rc_AA859980_at | - | - |
| Rat Genome U34 Set | rc_AA859980_g_at | - | - |
| Rat Genome U34 Set | H34047_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22289437 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361128 | - | - |
| RatRef-12 Expression BeadChip | ILMN_48603 | - | - |

Acat2
Genomic Map