Assay Details
| Gene Symbol: Sytl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127560.1 | NP_001121032.1 | 13 - 14 | 1704 | 66 | |
| XM_006227888.2 | XP_006227950.1 | 14 - 15 | 2260 | 66 | ||
| XM_006227891.2 | XP_006227953.1 | 14 - 15 | 2257 | 66 | ||
| GenBank mRNA | BC166706.1 | - | 13 - 14 | 1704 | 5599231 | 66 |
More Information
Updated Mapping Information
Set Membership: |
|

Sytl3
Genomic Map