Assay Details
| Gene Symbol: Cdk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108617.1 | NP_001102087.1 | 10 - 11 | 1253 | 71 | |
| XM_008762706.1 | XP_008760928.1 | 12 - 13 | 1739 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdk14
Genomic Map