Assay Details
| Gene Symbol: Gatad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006236076.2 | XP_006236138.1 | 1 - 2 | 562 | 104 | |
| XM_575359.4 | XP_575359.3 | 1 - 2 | 562 | 104 | ||
| GenBank mRNA | BC088430.1 | - | 2 - 3 | 161 | 104 | |
More Information
Set Membership: |
|

Gatad1
Genomic Map