Assay Details
| Gene Symbol: Gatad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006236076.2 | XP_006236138.1 | 3 - 4 | 757 | 114 | |
| XM_575359.4 | XP_575359.3 | 3 - 4 | 757 | 114 | ||
| GenBank mRNA | BC088430.1 | - | 4 - 5 | 356 | 114 | |
More Information
Set Membership: |
|

Gatad1
Genomic Map