Assay Details
| Gene Symbol: Bloc1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105941.2 | NP_001099411.2 | 3 - 4 | 348 | 65 | |
| GenBank mRNA | CB613801.1 | - | 3 - 4 | 354 | 65 | |
| FQ217708.1 | - | 3 - 4 | 361 | 65 | ||
| FQ221841.1 | - | 3 - 4 | 308 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bloc1s1
Genomic Map