Assay Details
| Gene Symbol: Plin5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134637.1 | NP_001128109.1 | 7 - 8 | 918 | 87 | |
| XM_008766770.1 | XP_008764992.1 | 8 - 9 | 925 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plin5
Genomic Map