Assay Details
| Gene Symbol: Mpnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085406.1 | NP_001078875.1 | 8 - 9 | 976 | 53 | |
| XM_008766773.1 | XP_008764995.1 | 9 - 10 | 1264 | 53 | ||
| GenBank mRNA | BC101893.1 | - | 7 - 8 | 804 | 53 | |
| BC158552.1 | - | 8 - 9 | 976 | 9098910 | 53 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mpnd
Genomic Map