Assay Details
Gene Symbol: Pot1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001073777.4 | XP_001073777.2 | 11 - 12 | 1528 | 135 | |
XM_006244268.2 | XP_006244330.1 | 12 - 13 | 1594 | 135 | ||
XM_006244269.2 | XP_006244331.1 | 12 - 13 | 1823 | 135 | ||
XM_006244270.2 | XP_006244332.1 | 12 - 13 | 2108 | 135 | ||
XM_006244271.2 | XP_006244333.1 | 12 - 13 | 2120 | 135 | ||
XM_008766793.1 | XP_008765015.1 | 11 - 12 | 1953 | 135 | ||
XM_008766794.1 | XP_008765016.1 | 8 - 9 | 2212 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
