Assay Details
Gene Symbol: Sult1c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031732.2 | NP_113920.2 | 6 - 7 | 737 | 159 | |
GenBank mRNA | BC088125.1 | - | 6 - 7 | 737 | 7324981 | 159 |
FQ219411.1 | - | 6 - 7 | 767 | 159 | ||
L22339.1 | - | 5 - 6 | 647 | 159 |
Gene Symbol: LOC100910235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244237.2 | XP_006244299.1 | 654 | 159 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
