Assay Details
Gene Symbol: Syp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012664.2 | NP_036796.1 | 1 | 86 | 61 | |
GenBank mRNA | BC099798.1 | - | 101 | 7936715 | 61 | |
X06177.1 | - | 95 | 61 | |||
X06388.1 | - | 127 | 61 | |||
X06655.1 | - | 59 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
