Assay Details
| Gene Symbol: Pls3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031084.1 | NP_112346.1 | 13 - 14 | 1592 | 75 | |
| XM_006257434.2 | XP_006257496.1 | 14 - 15 | 1727 | 75 | ||
| XM_006257438.2 | XP_006257500.1 | 13 - 14 | 2265 | 75 | ||
| XM_008773445.1 | XP_008771667.1 | 13 - 14 | 2444 | 75 | ||
| GenBank mRNA | BC092611.1 | - | 9 - 10 | 1059 | 75 | |
| X70706.1 | - | 13 - 14 | 1576 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P198716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580596 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835320 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826823 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378171 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125157 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P198716 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P198716 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P198716 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P833626 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P198716 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98492 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41718 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10981 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10937271 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937263 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937262 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937256 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937264 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937270 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937257 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937267 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937268 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937258 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937269 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937259 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937266 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937265 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937255 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937260 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937261 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10937254 | - | - |
| Rat Genome 230 2.0 Array | 1372947_at | - | - |
| Rat Genome 230 2.0 Array | 1371139_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371139_at | - | - |
| Rat Genome 230 2.0 Array | 1372947_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372947_at | - | - |
| Affymetrix HT Rat Focus Array | 1372947_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371139_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371139_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0X00006253.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006249.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006240.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006241.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006252.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006232.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006258.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006251.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006256.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006244.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006250.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006245.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006242.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006238.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006255.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006248.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006239.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006254.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006246.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00006257.rn.1 | TC0X00001017.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372947_PM_at | - | - |
| Rat Expression Set 230 | 1371139_at | - | - |
| Rat Expression Set 230 | 1372947_at | - | - |
| Rat Expression Set 230 | 1371139_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10937254 | - | - |
| Rat Genome U34 Set | X70706cds_at | - | - |
| Rat Genome U34 Set | rc_AI236227_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358347 | - | - |

Pls3
Genomic Map