Assay Details
| Gene Symbol: Nxt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773417.1 | XP_008771639.1 | 1 - 2 | 171 | 130 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008773417.1 transcripts.
Set Membership: |
|

Nxt2
Genomic Map