Assay Details
| Gene Symbol: Nxt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108120.1 | NP_001101590.1 | 2 - 3 | 267 | 92 | |
| XM_006257360.2 | XP_006257422.1 | 3 - 4 | 348 | 92 | ||
| XM_006257361.2 | XP_006257423.1 | 3 - 4 | 291 | 92 | ||
| XM_008773417.1 | XP_008771639.1 | 2 - 3 | 266 | 92 | ||
| GenBank mRNA | BC166561.1 | - | 2 - 3 | 203 | 7315205 | 92 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nxt2
Genomic Map