Assay Details
| Gene Symbol: Nxt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108120.1 | NP_001101590.1 | 1 - 2 | 176 | 77 | |
| XM_006257360.2 | XP_006257422.1 | 2 - 3 | 257 | 77 | ||
| XM_006257361.2 | XP_006257423.1 | 2 - 3 | 200 | 77 | ||
| GenBank mRNA | BC166561.1 | - | 1 - 2 | 112 | 7315205 | 77 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC166561.1, NM_001108120.1 transcripts.
Set Membership: |
|

Nxt2
Genomic Map