Assay Details
| Gene Symbol: Bmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191856.2 | NP_001178785.1 | 2 - 3 | 1036 | 86 | |
| GenBank mRNA | AF100787.1 | - | 2 - 3 | 281 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bmp7
Genomic Map