Assay Details
| Gene Symbol: Bmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191856.2 | NP_001178785.1 | 4 - 5 | 1383 | 87 | |
| GenBank mRNA | D29769.1 | - | 1 - 2 | 134 | 87 | |
More Information
Set Membership: |
|

Bmp7
Genomic Map