Assay Details
| Gene Symbol: Dok5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109344.2 | NP_001102814.1 | 2 - 3 | 254 | 67 | |
| XM_006235673.2 | XP_006235735.1 | 2 - 3 | 433 | 67 | ||
| GenBank mRNA | BC167027.1 | - | 2 - 3 | 254 | 7315978 | 67 |
More Information
Updated Mapping Information
Set Membership: |
|

Dok5
Genomic Map