Assay Details
Gene Symbol: Ccdc132 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173511.1 | NP_001166982.1 | 24 - 25 | 2395 | 105 | |
XM_006236037.2 | XP_006236099.1 | 24 - 25 | 2426 | 105 | ||
XM_008762704.1 | XP_008760926.1 | 24 - 25 | 2429 | 105 | ||
GenBank mRNA | BC091288.1 | - | 9 - 10 | 990 | 105 |
Gene Symbol: LOC100911994 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749697.3 | XP_003749745.2 | 4 - 5 | 1653 | 105 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
