Assay Details
| Gene Symbol: Lactb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106833.1 | NP_001100303.1 | 2 - 3 | 512 | 112 | |
| XM_008766242.1 | XP_008764464.1 | 2 - 3 | 495 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lactb
Genomic Map