Assay Details
| Gene Symbol: Cgnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108164.1 | NP_001101634.1 | 12 - 13 | 3168 | 59 | |
| XM_006243355.1 | XP_006243417.1 | 12 - 13 | 3112 | 59 | ||
| XM_008766286.1 | XP_008764508.1 | 12 - 13 | 3513 | 59 | ||
| XM_008766287.1 | XP_008764509.1 | 12 - 13 | 3168 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P144602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210716 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78763 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P144602 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P144602 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P144602 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P265333 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75028 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 54857 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10918614 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918608 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918607 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918604 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918602 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918611 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918606 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918615 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918601 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918613 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918612 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918610 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918616 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918618 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918605 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918609 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918617 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918603 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10918600 | - | - |
| Rat Genome 230 2.0 Array | 1391021_at | - | - |
| Rat Genome 230 2.0 Array | 1389107_at | - | - |
| Rat Genome 230 2.0 Array | 1391021_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389107_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391021_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391021_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800023320.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023316.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023326.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023315.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023325.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023311.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023310.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023323.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023327.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023328.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023324.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023321.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023312.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023317.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023309.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023319.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023322.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023305.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023307.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023313.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023329.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023318.rn.1 | TC0800002881.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0800023304.rn.1 | TC0800002881.rn.1 | RTA TC |
| Rat Expression Set 230 | 1389107_at | - | - |
| Rat Expression Set 230 | 1389107_PM_at | - | - |
| Rat Expression Set 230 | 1391021_at | - | - |
| Rat Expression Set 230 | 1391021_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10918600 | - | - |
| Rat Genome U34 Set | rc_AI008974_at | - | - |
| Rat Genome U34 Set | rc_AI059450_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364376 | - | - |

Cgnl1
Genomic Map